BBL™ BiGGY Agar, BD
Supplier: BD CANADA MICROBIOLOGY CA
211027
CA90000-046EA 229.29 CAD
CA90000-046
BBL™ BiGGY Agar, BD
Media for Microbiology Dehydrated Media for Microbiology
BiGGY (Bismuth Sulfite Glucose Glycine Yeast) is a selective and differential medium used in the detection, isolation, and presumptive identification of Candida species.
  • Differentiation is based on growth patterns and pigmentation
  • Bismuth sulfite acts as an inhibitory agent
  • Dextrose and yeast extract provide nutrients
  • Candida species produce brown to black pigmented colonies
Candida species reduce the bismuth sulfite, resulting in pigmentation of colonies and, with some species, pigmentation in the surrounding medium.