BBL™ Brain Heart Infusion, BD Biosciences
Supplier: BD CANADA MICROBIOLOGY CA
211059 299070
CA90000-060EA 202.23 CAD
CA90000-060 CA90001-530 CA90005-160
BBL™ Brain Heart Infusion, BD Biosciences
Media for Microbiology Dehydrated Media for Microbiology
Brain Heart Infusion (BHI) broth is a general-purpose liquid medium for the growth of a wide variety of bacterial and fungal species.
  • Brain and heart infusions and peptones provide nutrients
  • Supports the growth of fastidious and nonfastidious microorganisms
BHI Broth is used for the cultivation of a wide variety of microorganisms, including bacteria, yeasts, and molds.
Brain heart infusion, modified differs from other formulations by the quantities of the ingredients and the substitution of pancreatic digest of casein for pancreatic digest of gelatin.