Difco™ Baird-Parker Agar Base, BD
Supplier: BD CANADA MICROBIOLOGY CA
276810 276840
CA90003-972EA 2375.53 CAD
CA90003-972 CA90003-974
Difco™ Baird-Parker Agar Base, BD
Media for Microbiology Dehydrated Media for Microbiology
Baird-Parker Agar Base is used with EY (Egg Yolk) Tellurite Enrichment in the preparation of Egg-Tellurite-Glycerine-Pyruvate Agar (ETGPA) for selective isolation and enumeration of coagulase-positive staphylococci from food, skin, soil, air, and other materials.
  • Peptone, beef extract, and yeast extract provide essential nutrients
  • Sodium pyruvate stimulates growth of S. aureus
  • Egg yolk aids identification by demonstrating lecithinase activity
  • Glycine and lithium chloride inhibit growth of contaminant organisms
Baird-Parker Agar Base may also be used for identification of staphylococci on the basis of their ability to clear egg yolk.