Pseudomonas Agar F, BD
Supplier: BD CANADA MICROBIOLOGY CA
244820 244910
CA90003-352EA 360.05 CAD
CA90003-352 CA90003-354
Pseudomonas Agar F, BD
Media for Microbiology Dehydrated Media for Microbiology
Pseudomonas Agar F, also known as Flo Agar, is used for the enhancement of fluorescin production and Pseudomonas Agar P, also known as Tech Agar, is used for the enhancement of pyocyanin production by Pseudomonas.
  • 1:1 ratio of casein to meat peptone promotes fluorescin production
  • Magnesium sulfate provides ions for fluorescin production
  • Enzymatic digest of gelatin provides nitrogen and amino acids
  • Magnesium, potassium, and sulfate ions promote pyocyanin production
Both media contain glycerol, which acts as a source of energy and enhances pigment production.