Difco™ Potato Dextrose Media, BD
Supplier: BD CANADA MICROBIOLOGY CA
213400 213200 254920
CA90000-758EA 227.67 CAD
CA90000-758 CA90000-750 CA90000-754 CA90003-494
Difco™ Potato Dextrose Media, BD
Media for Microbiology Dehydrated Media for Microbiology
Potato Dextrose Agar is used for the cultivation and enumeration of yeasts and molds.
  • Potato starch and dextrose support luxuriant growth of fungi
  • Lowering the pH of the medium inhibits bacterial growth
  • Recommended media for testing nonsterile pharmaceutical products
Potato Dextrose Agar is a general-purpose medium for yeasts and molds that can be supplemented with acid or antibiotics to inhibit bacterial growth.
Potato Dextrose Broth is a general-purpose broth medium for cultivating yeasts and molds.