BBL™ Phenol Red Broth Base, BD Biosciences
Supplier: BD CANADA MICROBIOLOGY CA
211506 211514 211519
CA90000-208EA 237.64 CAD
CA90000-208 CA90000-210 CA90000-212
BBL™ Phenol Red Broth Base, BD Biosciences
Media for Microbiology Dehydrated Media for Microbiology
Phenol Red Broth Base and Phenol Red Broth with carbohydrates are used for the determination of fermentation reactions in the differentiation of microorganisms.
  • Pancreatic digest of casein provides nutrients
  • Phenol red acts as a pH indicator
  • Acid production causes a color change
Phenol Red Broth Base is a complete medium without added carbohydrate. It is used as a negative control for fermentation studies or as a base for the addition of carbohydrates.