Difco™ Oxford Medium, BD Diagnostics
Supplier: BD CANADA MICROBIOLOGY CA
222510
222530
222510
CA90001-650EA
1461.4
CAD
CA90001-650
CA90001-654
90001-650
Difco™ Oxford Medium, BD Diagnostics
Media for Microbiology
Dehydrated Media for Microbiology
Oxford Medium Base is used to prepare Oxford Medium or Modified Oxford Medium for isolating and differentiating Listeria monocytogenes.
- Listeria species grow over a pH range of 4.4-9.6
- Dehydrated appearance: Tan, free-flowing, homogeneous
- Motility is most pronounced at 20°C
The most common contaminating bacteria found in food sources potentially containing Listeria are streptococci, especially in the enterococci, micrococci and Bacillus species, Escherichia coli, Pseudomonas aeruginosa, and Proteus vulgaris.