Difco™ Nutrient Broth, BD
Supplier: BD CANADA MICROBIOLOGY CA
232000
231000
234000
CA90002-592EA
6138.54
CAD
CA90002-592
CA90002-108
CA90002-622
CA90002-660
Difco™ Nutrient Broth, BD
Media for Microbiology
Dehydrated Media for Microbiology
Nutrient Broth is used for the cultivation of many species of nonfastidious microorganisms.
- Beef extract provides carbohydrates, vitamins, nitrogen, and salts
- Peptones provide a source of nitrogen
Nutrient Broth is one of several nonselective media useful in routine cultivation of microorganisms.