Difco™ Nutrient Agar, BD
Supplier: BD CANADA MICROBIOLOGY CA
213000 211665
CA90000-744EA 301.61 CAD
CA90000-744 CA90000-260 CA90000-476
Difco™ Nutrient Agar, BD
Media for Microbiology Dehydrated Media for Microbiology
Nutrient Agar is used for the cultivation of bacteria and for the enumeration of organisms in water, sewage, feces, and other materials.
  • Beef extract provides carbohydrates, vitamins, nitrogen, and salts
  • Peptones provide a source of nitrogen
  • Agar is the solidifying agent
Nutrient Agar consists of peptone, beef extract, and agar. This relatively simple formulation provides the nutrients necessary for the replication of a large number of microorganisms that are not excessively fastidious.