TransPorter® Transport Swab, Liquid Amies
Supplier: Hardy Diagnostics
TransPorter®
4140BX
75832-482EA 91.99 CAD
75832-482
TransPorter® Transport Swab, Liquid Amies
Swabs Transport Swabs
Transport swabs with Amies liquid are designed for general microbiology and plate count applications.
  • Sterile
  • Contains no agar
A single, soft rayon swab on a plastic shaft with a round bottom plastic tube containing liquid modified Amies transport medium for general microbiology and plate count purposes. Ideally suited for throat, vaginal, wound, and skin swab applications. The 'Venturi' (hourglass) design locks in the sponge and specimen, and prevents exposure to air. Packaged in a clear oxygen barrier wrapper with a tamper-evident seal.