Difco™ Tryptic Soy Agar, BD Biosciences
Supplier: BD CANADA MICROBIOLOGY CA
236950 236920
CA90002-706EA 200.64 CAD
CA90002-706 CA90002-700 CA90002-704
Difco™ Tryptic Soy Agar, BD Biosciences
Media for Microbiology Additives and Supplements for Microbiology
Tryptic Soy Agar is used for the isolation and cultivation of nonfastidious and fastidious microorganisms.
  • Casein and soy peptones promote general growth
  • Sodium chloride maintains the osmotic equilibrium
Tryptic Soy Agar is not the medium of choice for anaerobes.