Difco™ Tinsdale Base, BD Biosciences
Supplier: BD CANADA MICROBIOLOGY CA
234210 278610
CA90002-668PK 1337.68 CAD
CA90002-668 CA90003-990
Difco™ Tinsdale Base, BD Biosciences
Media for Microbiology Additives and Supplements for Microbiology
Tinsdale Agar base is used with Tinsdale Enrichment, Desiccated in isolating and differentiating Corynebacterium diphtheriae.
  • Peptone provides nitrogen, vitamins, carbon, and amino acids
  • Sodium chloride maintains the osmotic balance of the medium
  • Agar is the solidifying agent
Tinsdale Enrichment, Dessicated contains bovine serum and horse serum, which provide essential growth factors.