BBL™ Thioglycollate Medium without Indicator, BD Life Sciences
Supplier: BD CANADA MICROBIOLOGY CA
211720 221199
CA90000-298EA 267.94 CAD
CA90000-298 CA90001-262
BBL™ Thioglycollate Medium without Indicator, BD Life Sciences
Media for Microbiology Additives and Supplements for Microbiology
Thioglycollate Medium without Indicator is a general-purpose medium for the cultivation of microorganisms, especially obligate anaerobes.
  • Casein and soybean peptones, dextrose, and cystine supply nitrogen and carbon
  • Sodium chloride provides essential ions
Casein and soybean peptones, dextrose, and cystine supply nitrogenous and carbonaceous compounds, fermentable carbohydrates, and trace ingredients. This medium is satisfactorily used as an enrichment culture for various types of specimens and also as a transportation medium.