Difco™ Supplement VX with Reconstituting Fluid, BD Biosciences
Supplier: BD CANADA MICROBIOLOGY CA
233541 233542
CA90002-650PK 697.13 CAD
CA90002-650 CA90002-652
Difco™ Supplement VX with Reconstituting Fluid, BD Biosciences
Media for Microbiology Additives and Supplements for Microbiology
Supplement VX with Reconstituting Fluid is used for supplementing media to culture fastidious microorganisms, particularly Neisseria gonorrhoeae and Haemophilus influenzae.
  • Lyophilized concentrate
  • Contains vitamins, amino acids, coenzymes, and dextrose
This supplement is recommended for enriching GC Agar media, Proteose No. 3 Agar, and the selective agars for the isolation of pathogenic Neisseria.