Difco™ Tryptone Glucose Yeast Agar (Standard Methods Agar), BD Life Sciences
Supplier: BD CANADA MICROBIOLOGY CA
247910
247940
CA90003-404EA
916.91
CAD
CA90003-404
CA90003-408
CA90003-410
Difco™ Tryptone Glucose Yeast Agar (Standard Methods Agar), BD Life Sciences
Media for Microbiology
Dehydrated Media for Microbiology
Tryptone Glucose Yeast Extract Agar is recommended for the enumeration of bacteria in water, air, milk, and dairy products.
- Glucose is the energy source
- Dehydrated appearance: Cream to yellow, free-flowing
- Firm, comparable with 1.5% Agar gel
Tryptone Glucose Yeast Extract Agar is used as a standard medium for the bacteriological plate count of milk and dairy products.