Difco™ Sabouraud Dextrose Agar, BD
Supplier: BD CANADA MICROBIOLOGY CA
210950 211661
CA90000-039EA 230.45 CAD
CA90000-039 CA90000-258
Difco™ Sabouraud Dextrose Agar, BD
Media for Microbiology Dehydrated Media for Microbiology
Sabouraud Dextrose Agar is used in qualitative procedures for the cultivation of dermatophytes and other pathogenic and nonpathogenic fungi from clinical and nonclinical specimens.
  • Low pH is favorable for the growth of fungi
  • Dehydrated appearance: Light beige, free-flowing
  • Peptones are the source of nitrogenous growth factors
The addition of antimicrobics further increases the selectivity of the medium.