Riboflavin Assay Medium, BD
Supplier: BD CANADA MICROBIOLOGY CA
232510
232510
CA90002-608EA
165.39
CAD
CA90002-608
90002-608
Riboflavin Assay Medium, BD
Media for Microbiology
Dehydrated Media for Microbiology
Riboflavin Assay Medium is used for determining riboflavin concentration by the microbiological assay technique.
- Dehydrated appearance: Beige, free-flowing
- Free of riboflavin but contains all other nutrients and vitamins essential for the growth of Lactobacillus rhamnosus
The addition of riboflavin in specified increasming concentrations gives a growth response that can be measured turbidimetrically or titrimetrically.