Difco™ Rhamnose, BD
Supplier: BD CANADA MICROBIOLOGY CA
217510 217520
CA90000-942EA 369.03 CAD
CA90000-942 CA90000-944
Difco™ Rhamnose, BD
Media for Microbiology Dehydrated Media for Microbiology
Rhamnose media are used to differentiate bacteria on the basis of carbohydrate fermentation abilities.
  • Naturally occurring deoxy sugar
  • Commonly binds to other sugars in nature
Rhamnose can be isolated from Buckthorn, poison sumac, and plants in the genus Uncaria, and is also produced by microalgae belonging to class Bacillariophyceae.