Rappaport Vassiliadis Salmonella (RVS) Soy Broth, BD Diagnostics
Supplier: BD CANADA MICROBIOLOGY CA
215199
214943
CA95038-632PK
75.23
CAD
CA95038-632
CA95038-637
Rappaport Vassiliadis Salmonella (RVS) Soy Broth, BD Diagnostics
Media for Microbiology
Dehydrated Media for Microbiology
Rappaport-Vassiliadis Salmonella Soy Broth is used for selectively enriching Salmonella in food and environmental samples.
- Soy peptone is a source of carbon and nitrogen for general growth requirements
- Malachite green helps inhibit organisms other than Salmonella
- Low pH helps select for highly resistant Salmonella species
Meets United States Pharmacopeia (USP), European Pharmacopoeia (EP), and Japanese Pharmacopoeia (JP) performance specifications, where applicable.