BBL™ Listeria Selective Supplement, BD Biosciences
Supplier: BD CANADA MICROBIOLOGY CA
212402
CA90000-664PK 625.16 CAD
CA90000-664
BBL™ Listeria Selective Supplement, BD Biosciences
Media for Microbiology Dehydrated Media for Microbiology
Listeria Selective Supplement is an antimicrobial supplement recommended for the selective isolation and identification of Listeria monocytogenes.
  • Selective isolation
  • Differentiation
  • Dispensed aseptically
Listeria Selective Supplement is used for the detection of Listeria monocytogenes.