BBL™ Columbia CNA Agar Dehydrated Media, BD Diagnostics
Supplier: BD CANADA MICROBIOLOGY CA
212104 294221
CA90000-486EA 250.45 CAD
CA90000-486 CA90004-542
BBL™ Columbia CNA Agar Dehydrated Media, BD Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Columbia CNA Agar is used with blood for selective isolation of Gram-positive cocci.
  • Selective medium
  • Contains two antibiotics that inhibits the growth of Gram-negative bacteria
  • Contains whole blood to allow for differentiation based on hemolysis patterns
Columbia CNA Agar is recommended for use as a selective growth medium for the isolation and differentiation of Gram-positive cocci from clinical and non-clinical specimens which contain mixed flora.