Inoculum Broth, BD
Supplier: BD CANADA MICROBIOLOGY CA
211813 254210
CA90000-366EA 1269.54 CAD
CA90000-366 CA90003-490
Inoculum Broth, BD
Media for Microbiology Dehydrated Media for Microbiology
Inoculum Broth is used for preparing the inoculum of lactobacilli and other microorganisms used in microbiological assays of vitamins and amino acids.
  • Used for cultivating lactobacilli
  • For preparing the inoculum for microbiological assays
  • Peptone provides nitrogen and amino acids
  • Beige, homogenous appearance when dehydrated
Inoculum Broth is the recommended medium for preparation of lactobacilli inoculum for vitamin assays.
Order Now
SPECIFICATIONS
Environmentally Preferable