Difco™ Salmonella O Antigen and Antisera Groups, BD
Supplier: BD CANADA MICROBIOLOGY CA
211778 221301 221421 222591
CA90000-326EA 675.84 CAD
CA90000-326 CA90000-350 CA90001-294 CA90001-302 CA90001-370 CA90001-662
Difco™ Salmonella O Antigen and Antisera Groups, BD
Media for Microbiology Dehydrated Media for Microbiology
BD Difco™ QC Antigens Salmonella O and BD Difco QC Antigen Salmonella Vi are used in the quality control testing of BD Difco Salmonella O Antisera and BD Difco Salmonella Vi Antiserum by slide agglutination tests.
  • May be used to demonstrate a negative control reaction
  • Chemically stable
  • Inactivated suspension
These antigens are chemically stabilized and inactivated suspensions of known strains of the genus Salmonella designed for use as homologous controls for testing the efficacy of the Salmonella typing antisera employed in routine laboratory procedures.