BBL™ Trichosel™ Broth, BD Biosciences
Supplier: BD CANADA MICROBIOLOGY CA
211747
298323
CA90000-320EA
729.75
CAD
CA90000-320
CA90005-126
BBL™ Trichosel™ Broth, BD Biosciences
Media for Microbiology
Dehydrated Media for Microbiology
BBL™ Trichosel™ Broth is used for the isolation and cultivation of Trichomonas species.
- Modification of the STS Medium of Kupferberg et al.
- For the cultivation of Trichomonas
- Contains casein peptone, cysteine, beef extract, and yeast extract
Chloramphenicol is a broad-spectrum antibiotic which inhibits a wide range of Gram-positive and Gram-negative bacteria. Horse serum is added to provide growth factors required by Trichomonas.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...