BBL™ Fluid Thioglycollate, BD
Supplier: BD CANADA MICROBIOLOGY CA
221196
299108
CA-90001-260PK
354.39
CAD
CA-90001-260
CA90001-144
CA90000-132
CA90001-258
CA90005-176
BBL™ Fluid Thioglycollate, BD
Media for Microbiology
Dehydrated Media for Microbiology
Fluid thioglycolate is a reducing medium containing sodium thioglycolate, which reacts with molecular oxygen to keep free oxygen levels low.
- Presence creates a redox potential in test tubes
- Allows the differentiation of various aerobes and anaerobes
Fluid thioglycolate medium is a multipurpose, enriched, differential medium used primarily to determine the oxygen requirements of microorganisms.