BBL™ MGIT™ Mycobacteria Growth Indicator, BD Diagnostics
Supplier: BD CANADA MICROBIOLOGY CA
245111 245113 245114 245116
CA-90003-368PK 840.98 CAD
CA-90003-368 CA-90003-370 CA-90003-372 CA-90003-374
BBL™ MGIT™ Mycobacteria Growth Indicator, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
The BBL™ Mycobacteria Growth Indicator Tube is a rapid, easy-to-use system with high accuracy for the detection of mycobacteria directly from clinical specimens.
  • Growth in as little as four to six days
  • Unmistakable results
  • Outstanding performance
  • Fits perfectly into your laboratory work flow
The BBL™ MGIT™ Mycobacteria Growth Indicator Tube supplemented with BBL™ MGIT™ OADC enrichment and BBL™ MGIT™ PANTA™ antibiotic mixture, when appropriate, is intended for the detection and recovery of mycobacteria.
Acceptable specimen types are digested and decontaminated clinical specimens (except urine) and sterile body fluids (except blood).
BBL™ MGIT™ tubes also fit into the standard procedures for decontamination (NaCl-NaOH) and disposal of waste. It is ideal for any size laboratory, especially those performing a high volume of mycobacteria tests.