AmnioTest™, Pro-Lab Diagnostics
Supplier: PRO-LAB DIAGNOSTICS INC CA
PL.901 PL.901-20 PL.902 PL.910 PL.911 PL.912 PL.913 PL.914 PL.915
75784-944BX 669.97 CAD
75784-944 75784-946 75784-948 75784-950 75784-952 75784-954 75784-956 75784-958 75784-960
AmnioTest™, Pro-Lab Diagnostics
Clinical Diagnostic Supplies Clinical Reagents and Kits
AmnioTest™ is a qualitative, pH-based, swab screening system intended as an aid in detecting rupture of amniotic membrane in pregnant women.
  • Detect rupture of amniotic membrane in pregnant women
  • Determine the presence of amniotic fluid
  • Swab absorbs fluid associated with the tissue and the dye develops a colour which correlates with the pH of the absorbed fluid
Rupture of the amniotic membrane can result in small volumes of amniotic fluid leaking into the upper vagina
A swab impregnated with nitrazine yellow dye is brought into contact with the upper vagina. The swab absorbs fluid associated with the tissue and the dye develops a colour which correlates with the pH of the absorbed fluid over the range pH 5.0 to pH 7.5. Amniotic fluid has a neutral pH while the pH of the upper vagina is normally acidic. A pH of 6.5 or higher in the upper vagina is consistent with leakage of amniotic fluid.
The presence of amniotic fluid tends to elevate the pH of the upper vagina. Detection of this pH increase using a pH indicator dye has been shown to assist in determining the presence of amniotic fluid.
Order Now
SPECIFICATIONS
Environmentally Preferable