Letheen Broth, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
Letheen Broth is used for determining the phenol coefficient of cationic surface-active materials.
- Determine the antimicrobial activity of quaternary ammonium compounds
- Microbial evaluation of personal care products
- For sample preparation and dilution of cosmetics and personal care products
Recommended for use to determine the antimicrobial activity of quaternary ammonium compounds and in the microbial evaluation of personal care products.
For sample preparation and dilution of cosmetics and personal care products for microbiological examination and for the bactericidal evaluation of quaternary ammonium disinfectants.
Ordering information: Minimum Order of 10 packs of 10.
Caution: *Product not to be used clinically or for patient reporting.