Mycobiotic Agar Slant, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
W50
L45
75814-674PK
61.23
CAD
75814-674
75815-004
Mycobiotic Agar Slant, Hardy Diagnostics
Media for Microbiology
Dehydrated Media for Microbiology
Mycobiotic Agar Slant is used for the selective isolation of pathogenic fungi.
- Selective isolation of pathogenic fungi
- Contains chloramphenicol and cyclohexamide.
For the selective isolation of pathogenic fungi especially from hair, skin, and nail specimens. Contains chloramphenicol and cyclohexamide.