Human Beta-Amyloid (1-42), FAM (Carboxyfluorescein)
Supplier: Anaspec Inc
This is a fluorescent (FAM)-labeled ß-Amyloid peptide, Abs/Em=494/521 nm.
Sequence: FAM-DAEFRHDSGYEVHHQKLVFFAEDVGSNKGAIIGLMVGGVVIA
MW: 4872.4 Da
% Peak Area by HPLC: ≥95
Peptide Content: ≥ 60%
Storage condition: -20°C
Citation:
Fleisher-Berkovich, S. et al. (2010). Distinct modulation of microglial amyloid β phagocytosis and migration by neuropeptides. J Neuroinflammation 7, 61. doi: 10.1186/1742-2094-7-61
Choucair, A. et al. (2007). Preferential accumulation of Aβ(1-42) on gel phase domains of lipid bilayers: An AFM and fluorescence study. Biochim Biophys Acta-Biomembranes 1768, 146.
Saavedra, L. et al. (2007). Internalization of β-amyloid peptide by primary neurons in the absence of apolipoprotein E. J Biol Chem 282, 35722.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...