Brain Heart Infusion Agar, Life Technologies
Supplier: BD CANADA MICROBIOLOGY CA
221610 295756
CA90000-508EA 1186 CAD
CA90000-508 CA90001-402 CA90004-576
Brain Heart Infusion Agar, Life Technologies
Media for Microbiology Dehydrated Media for Microbiology
Brain Heart Infusion agar is a general-purpose medium suitable for the cultivation of a wide variety of organism types, including bacteria, yeasts, and molds.
  • General-purpose medium
  • Promotes growth of bacteria, yeasts, molds, and fungi
With the addition of 5% or 10% sheep blood, it is used for the isolation and cultivation of a wide variety of fungal species, including systemic fungi, from clinical and nonclinical sources.