MacConkey Agar, BD
Supplier: BD CANADA MICROBIOLOGY CA
211387
211391
CA90000-168EA
199.55
CAD
CA90000-168
CA90000-172
MacConkey Agar, BD
Media for Microbiology
Dehydrated Media for Microbiology
MacConkey agar is a selective and differential culture medium for bacteria designed to selectively isolate Gram-negative and enteric bacilli and differentiate them based on lactose fermentation.
- Isolates Gram-negative bacilli
Enteric bacteria that have the ability to ferment lactose can be detected using the carbohydrate lactose, and the pH indicator neutral red.