BBE/LKV (Bacteroides Bile Esculin/Laked Blood Kanamycin Vancomycin) Biplate, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
J102
J102
10324-540EA
75.46
CAD
10324-540
CA10001-454
BBE/LKV (Bacteroides Bile Esculin/Laked Blood Kanamycin Vancomycin) Biplate, Hardy Diagnostics
Media for Microbiology
Additives and Supplements for Microbiology
For the selective isolation of B
- Selective isolation of gram negative anaerobic bacteria
(Bacteroides Bile Esculin and Laked Blood with Kanamycin and Vancomycin Biplate) For the selective isolation of B. fragilis group and selective isolation of gram negative anaerobic bacteria.
fragilis group.