Rapid Ornithine, for Enterobacteriaceae and Staphylococcus Lugdunensis
Supplier: HARDY DIAGNOSTICS
K279
K279
10324-464EA
168.98
CAD
10324-464
CA10001-470
Rapid Ornithine, for Enterobacteriaceae and Staphylococcus Lugdunensis
Media for Microbiology
Dehydrated Media for Microbiology
This test will assist in the identification of Enterobacteriaceae and Staphylococcus lugdunesis.
- Detect ornithine decarboxylase activity in as little as two to four hours
- Assist in the identification of Enterobacteriaceae and Staphylococcus lugdunesis
Hardy Diagnostics Rapid Ornithine can be used to detect ornithine decarboxylase activity in as little as two to four hours. This test will assist in the identification of Enterobacteriaceae and Staphylococcus lugdunesis.