CatScreen™, Rapid Test for Moraxella (Branhamella) Catarrhalis (M. cat.), Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
Z110 Z110
10324-426EA 47.1 CAD
10324-426 CA10001-512
CatScreen™, Rapid Test for Moraxella (Branhamella) Catarrhalis (M. cat.), Hardy Diagnostics
Microbiology Test Systems Microorganism Tests
Rapid test for the detection of the enzyme butyrate esterase in bacteria.
  • Performs a butyrate test within five minutes
  • Presumptive identification of Branhamella (Moraxella) catarrhalis
  • Rapid, red color test result
Rapid test for the detection of the enzyme butyrate esterase in bacteria isolated on culture media for the presumptive identification of Branhamella (Moraxella) catarrhalis. Performs a butyrate test within five minutes. Simply moisten disk, inoculate with organism, then wait for the development of a red color.