Page's Saline, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
R225
R225
10324-386EA
168.14
CAD
10324-386
CA10001-492
Page's Saline, Hardy Diagnostics
Buffers
Cell Culture Buffers
Page's Saline for detecting the presence of free-living amebae cysts and trophozoites in tissue, soil, or water samples.
- For use with non-nutrient agar
- Simple procedure
- Microscopically visualize distinct feeding tracks
For use with Non-Nutrient Agar for detecting the presence of free-living amebae cysts and trophozoites in tissue, soil, or water samples.
Simple procedure, prepared, ready-to-use media. Easy Read-out, microscopically visualize (100x) distinct feeding tracks left behind by amebae.