CNA Agar, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
A50 J62
10063-766EA 43.26 CAD
10063-766 10324-550
CNA Agar, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CAN Agar is ideal for the culture and enumeration of urinary tract pathogens from clinical specimens.
  • Gram positive bacteria on the CNA side
  • Gram negative bacteria on the MAC side
  • Polystyrene Petri plates feature raised rings for stacking ability
Columbia Agar with 5% Sheep Blood, Colistin and Nalidixic Acid / MacConkey Agar is a two-section Petri plate of CNA/MAC
Contains 5% sheep blood from a veterinary supervised, controlled herd of donor animals. Blood is guaranteed to be from healthy antibiotic-free animals.
Polystyrene Petri plates feature raised rings for stacking ability and prevents sliding.
Ideal for the culture and enumeration of urinary tract pathogens from clinical specimens. Only Gram positive bacteria will grow on the CNA side. Only Gram negative bacteria will grow on the MAC side.