GBS Detect/Strep Reagents
Supplier: HARDY DIAGNOSTICS
A300 A70 G123 G123 Z75 A300BX
10324-334EA 68 CAD
10324-334 10324-342 10324-504 CA10001-426 CA10001-530 CA77623-656
GBS Detect/Strep Reagents
Microbiology Test Systems Microorganism Tests
GBS Detect is used for the isolation and differentiation by enhanced hemolysis of gamma hemolytic Group B Streptococcus.
  • Grouping reagents for the rapid latex agglutination test
  • Lancefield grouping includes A,B,C,D,F,G latex, controls, extraction reagents, cards, and mixing sticks
  • StrepQuick™ to aid in the identification of gram positive cocci
GBS Detect™, for non-hemolytic Group B Strep. Granada Medium for hemolytic Group B Strep. Grouping Reagents A, B, C, and G, for the rapid latex agglutination test. PYR Test Kit with chromogenic developer on paper disks. Selective Beta Strep Agar, for Group A Strep only (Streptococcus pyogenes), especially for throat cultures. Selective Strep Agar (COBA Medium), for the selective isolation of all species of Strep. ci. StrepQuick™ performs a PYR, LAP, and Esculin test to aid in the identification of gram positive coc.