Trio.Bas™ DUO Kits
Supplier: Hardy Diagnostics
BAS220 BAS221 BAS226 BAS220K BAS225K BAS221K BAS226K
75804-716EA 14439.19 CAD
75804-716 75804-718 75804-722 CA76049-412 CA76049-414 CA76168-944 CA76168-946
Trio.Bas™ DUO Kits
Samplers Air Samplers
Hardy Diagnostics introduces the next generation of microbial air samplers for viable particles.
  • Two aspirating heads with easy manipulation bayonet closure
  • Ability to use two different culture media at the same time
  • Save sampling time by doubling the aspirated volume of air
  • Manual or automatic operations
All of the air samplers are lightweight, with an antibacterial techno-polymer shockproof body. They have a long lasting battery and are IP65 protected from dust and water.
Per USP 797, 'Impaction is the preferred method of active air sampling. Use of settling plates for qualitative air sampling cannot be relied upon and shall not be used solely to determine the quality of air in the controlled environment.'
Certifications: ISO14698, USP 797, USP 116, EU GPL-GMP, ISO 17025.
Accessories information: Additional accessory components, including data transfer options, are available separately.
Delivery information: Trio.Bas™ DUO Kit Contains:
Trio.Bas™ DUO air sampler with Bluetooth data transfer, includes calibration certificate
Induction battery charger; unit simply rests on base station to charge
Stainless steel aspirating head (×2), autoclavable
Stainless steel cover head (×2), autoclavable
Robustus hard shell carrying case, to protect sampler in transit
IQ, OQ, PQ, fillable documents