Accessory Parts for TRIO.BAS™ Air Samplers
Supplier: Hardy Diagnostics
BAS395 BAS420 BAS421 BAS330 BAS331 BAS334 BAS336 BAS340 BAS341 BAS265 BAS465 BAS521 BAS522 BAS530 BAS371 BAS523 BAS524 BAS520 BAS197
75804-696EA 1161.21 CAD
75804-696 75804-700 75804-702 75804-704 75804-746 75804-748 75804-750 75804-754 75804-756 75804-758 75838-740 CA75994-444 CA75994-446 CA75994-448 CA75994-450 CA75994-452 CA75994-454 CA75994-464 CA75994-470 CA76509-286
Accessory Parts for TRIO.BAS™ Air Samplers
Samplers Air Samplers
An assortment of accessory and replacement products designed for use with the Hardy Diagnostics collection of TRIO.BAS™ air samplers.
Aspirating heads are available in stainless steel, Daily Shift polystyrene, or polycarbonate formats. The durable stainless steel and polycarbonate models are autoclavable and can be sprayed with disinfectant. The Daily Shift polystyrene models are supplied ready to use in a cleanroom environment. These heads are individually packaged with a Certificate of Sterility provided. The Blind Head is designed to protect the aspirating chamber when instrument is not in use.
Ordering information: Stainless steel aspirating head models BAS330 and BAS331 and induction battery charger model BAS318 are required when instruments are purchased a la carte instead of the kit format. Vertical hook and wall holder require knob (sold separately).