Trio.Bas™ MONO Kits
Supplier: Hardy Diagnostics
BAS200 BAS201 BAS205 BAS206 BAS200K BAS205K BAS201K BAS206K
75804-708EA 12387.12 CAD
75804-708 75804-710 75804-712 75804-714 CA76049-408 CA76049-410 CA76168-940 CA76168-942
Trio.Bas™ MONO Kits
Samplers Air Samplers
Hardy Diagnostics introduces the next generation of microbial air samplers for viable particles.
  • Aspirating head with easy manipulation bayonet closure
  • Manual or automatic operations
  • Ergonomic and balanced design to facilitate handling with ease
  • No plugs or external connections
All of the air samplers are lightweight, with an antibacterial techno-polymer shockproof body. They have a long lasting battery and are IP65 protected from dust and water
Per USP 797, 'Impaction is the preferred method of active air sampling. Use of settling plates forMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable