Pierce™ Imject™ Bovine Serum Albumin (BSA), Maleimide-activated
Supplier: Thermo Scientific
77115
77116
CAPI77115EA
2571.32
CAD
CAPI77115
CAPI77116
Pierce™ Imject™ Bovine Serum Albumin (BSA), Maleimide-activated
Proteins and Peptides
Thermo Scientific Imject Maleimide-Activated BSA enables conjugation of sulfhydryl-containing peptide antigens to immunogenic bovine serum albumin carrier protein for immunization and antibody production.
- Optimized activation
- Sulfhydryl-reactive
- Simple, rapid procedure
- High-quality carrier protein
- Immunogenic and soluble