Pierce Imject™ Mariculture KLH Carrier Proteins, Thermo Scientific
Supplier: Thermo Scientific
77600 77649 77653
CAPI77600EA 2335.75 CAD
CAPI77600 CAPI77649 CAPI77653
Pierce Imject™ Mariculture KLH Carrier Proteins, Thermo Scientific
Protein Extraction Protein Extraction Kits
Imject™ Mariculture KLH (mcKLH) is a purified keyhole limpet haemocyanin carrier protein that enables simple preparation of highly effective immunogens with peptide antigens.
  • High-yield conjugation with each molecule of KLH contains hundreds of primary amines available for coupling haptens via EDC or NHS ester crosslinkers
  • Validated quality with purified and stabilized mariculture KLH maintains solubility in aqueous solutions
  • Sustainable source with KLH that is harvested from select populations of the mollusc that are grown in mariculture
  • Highly immunogenic as KLH has a high molecular mass that elicits a stronger immune response
Mariculture KLH is purified and lyophilized in buffers that optimize its stability and solubility for hapten conjugation
Preparations in phosphate buffer (PBS) are ready for hapten-carrier conjugation via amine-reactive NHS-ester or glutaraldehyde crosslinking. Preparations in MES buffer are optimized for peptide-carrier conjugation via the carboxyl-reactive carbodiimide crosslinker called EDC (or EDAC).
The large, complex protein contains hundreds of sites per molecule for effective conjugation of peptides and other antigens using amine-reactive or carboxyl-reactive crosslinkers.