Urine Test Strips, Germaine Laboratories
Supplier: Germaine Laboratories
Reagent strips for the qualitative and semi-quantitative detection of various analytes in urine, up to 10
The MicroAlbumin 2-1 test detects Microalbumin in concentration levels as low as 10 mg/L and Creatinine at 100 mg/L (MicroAlbumin CPT Code 82044 QW, Creatinine CPT Code 82570 QW).
A color chart is provided on each bottle to easily read results.
Certifications: CLIA waived.
Ordering information: AimStrip® 10-SG and AimStrip® US-4OB tests are compatible with the Aimstrip® Urine Analyzer.