Anti-HSF1 Rabbit Polyclonal Antibody

Supplier: ProSci
79-830
10813-698EA 868.4 CAD
10813-698
Anti-HSF1 Rabbit Polyclonal Antibody
Antibodies
DNA-binding protein that specifically binds heat shock promoter elements (HSE) and activates transcription. In higher eukaryotes, HSF is unable to bind to the HSE unless the cells are heat shocked.

Type: Primary
Antigen: HSF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR