Modified Letheen Agar, BD Difco™, BD Life Sciences
Supplier: BD CANADA MICROBIOLOGY CA
263110 292847
CA90003-734EA 324.71 CAD
CA90003-734 CA90004-444
Modified Letheen Agar, BD Difco™, BD Life Sciences
Media for Microbiology Dehydrated Media for Microbiology
Modified letheen agar neutralizes quaternary ammonium compounds and is used for microbiological evaluation of cosmetics.
  • Neutralizes quaternary ammonium compounds
  • Microbiological evaluation of cosmetics