BD® Fluid Thioglycollate Medium, Special (ETO)l, BD Diagnostics
Supplier: BD CANADA MICROBIOLOGY CA
257097 257544 257097 257544 257548
10809-160PK 2152.69 CAD
10809-160 10809-724 CA10809-160 CA10809-724 76314-972
BD® Fluid Thioglycollate Medium, Special (ETO)l, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
Fluid Thioglycollate Medium is used for the sterility testing of biologics and for the enrichment and cultivatin of anaerobes, aerobes, and microaerophiles.
  • ETO (Ethylene Oxide) gassed bottles combine the advantages of standard bottles with additional features
  • Outer packaging sterilization removes the need for spraying down bottles before introducing them into cleanrooms or isolators
  • Polypropylene boxes reduce the shedding of particles often associated with cardboard that can cause issues with pharmaceutical manufacturing
  • Bottles are wrapped in a polypropylene box, a Stericlin® bag, and an outer transport box to ensure sterility assurance level (SAL) required by pharmaceutical users
  • Users no longer have to sterilize by autoclave before use, an important time and labor-saving step
  • Multiple bottles packed in the box allows for easy transfer of a high number of sterility testing bottles into clean rooms at one time
  • Screw cap with septum allows a variety of sterility testing methods to be used