BD® Tryptic Soy Broth (TSB), Ethylene Oxide (ETO), Bottles, BD Diagnostics
Supplier: BD CANADA MICROBIOLOGY CA
257307
257543
257307
257543
257547
10809-720PK
2181.76
CAD
10809-720
10809-722
CA10809-720
CA10809-722
76314-970
BD® Tryptic Soy Broth (TSB), Ethylene Oxide (ETO), Bottles, BD Diagnostics
Media for Microbiology
Prepared Media for Microbiology
Tryptic Soy Broth is a general purpose microbiology medium used in qualitative procedures for the detection, isolation, and cultivation of fastidious and non-fastidious microorganisms used in sterility testing.
- ETO (Ethylene Oxide) gassed bottles combine the advantages of standard bottles with additional features
- Outer packaging sterilization removes the need for spraying down bottles before introducing them into cleanrooms or isolators
- Polypropylene boxes reduce the shedding of particles often associated with cardboard that can cause issues with pharmaceutical manufacturing
- Bottles are wrapped in a polypropylene box, a Stericlin® bag, and an outer transport box to ensure sterility assurance level (SAL) required by pharmaceutical users
- Users no longer have to sterilize by autoclave before use, an important time and labor-saving step
- Multiple bottles packed in the box allows for easy transfer of a high number of sterility testing bottles into clean rooms at one time
- Screw cap with septum allows a variety of sterility testing methods to be used