Pierce™ TurboFect™ Transfection Reagent, Thermo Scientific
Supplier: Thermo Scientific
Thermo Scientific TurboFect™ transfection reagent is a highly efficient, easy-to-use, non-immunogenic transfection reagent for delivery of DNA plasmids and expression vectors into eukaryotic cells.
- High transfection efficiency of a wide variety of cell types
- No optimization required for most cell lines
- Excellent transfection efficiency in the presence or absence of serum
- Minimal cytotoxicity
- Ready-to-use without need to reconstitute, dilute or manipulate
- Store at 4 °C
- Simple protocol
It is a sterile solution of a proprietary cationic polymer which interacts with DNA to form small, stable, highly diffusible complexes which are readily endocytosed. TurboFect™ reagent is ideal for transfection of a variety of cells, including primary and difficult-to-transfect cells.
High transfection efficiency of plasmid DNA into a wide variety of cell types, that is adherent and suspension cells and differentiated and undifferentiated cells.
Caution: For Research Use Only. Not for use in diagnostic procedures.