RANTES ELISA Kit, Human, Thermo Scientific
Supplier: Thermo Scientific
Use the Thermo Scientific® Human RANTES ELISA Kit to assay RANTES (regulated upon activation, normal T-cell expressed and secreted) protein levels in human serum, plasma, tissue extracts, urine, and cell culture supernatant samples. This is an alternate product for Cat. Nos. KHC1031 and KAC1511.
This kit uses pre-titered, matched pairs of coating and detection antibodies to achieve sensitive and accurate measurement of RANTES protein levels in samples. The Thermo Scientific® Human RANTES ELISA Kit also includes calibrated recombinant protein standard, streptavidin-HRP, TMB substrate solution, and stop solution. This complete kit contains one pre-coated 96-well strip plate and lot-optimized reagents. All components undergo quality control testing to minimize lot-to-lot variability.
Highlights:
Target: natural and recombinant human RANTES (regulated upon activation, normal T-cell expressed and secreted) protein
Assay range: 51.2 to 2000 pg/mL
Ordering information: For Research Use Only. Not for use in diagnostic procedures. Pre-coated plate(s), standard or calibrator, detector antibody, HRP conjugate, diluents, wash buffer, substrate(s), and stop solution. Store kit at 2-8˚C. See product manual for detailed contents and storage conditions for maximum stability.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...